2,3,4-Trimethoxyphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19838-03
L19838-06
CAAAAL19838-03EA
40.92
CAD
CAAAAL19838-03
CAAAAL19838-06
2,3,4-Trimethoxyphenylboronic acid 98%
2,3,4-Trimethoxyphenylboronic acid
Purity: 98%
CAS No.: 118062-05-8
Molecular Formula: C9H13BO5
Formula Weight: 212.01
Storage Temperature: -30°C to -10°C
Melting Point: 74-79°
Beilstein Reference: 4185924
MDL No.: MFCD01318183
Formula:
C₉H₁₃BO₅ Melting Pt: 74…79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318183 CAS Number: 118062-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...