2,4,6-Trimethoxyphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4,6-Trimethoxybenzeneboronic acid, (2,4,6-Trimethoxyphenyl)metaboric acid, Dihydroxy(2,4,6-trimethoxyphenyl)borane

L19837-03 L19837-06
CAAAAL19837-03EA 139.2 CAD
CAAAAL19837-03 CAAAAL19837-06
2,4,6-Trimethoxyphenylboronic acid 98%
2,4,6-Trimethoxyphenylboronic acid

Purity: 98% CAS No.: 135159-25-0 Molecular Formula: C9H13BO5 Formula Weight: 212.01 Storage Temperature: -30°C to -10°C Melting Point: 96-100° Beilstein Reference: 4315581 MDL No.: MFCD01114642

Formula: C₉H₁₃BO₅
MW: 212.01 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01114642
CAS Number: 135159-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR