2-Amino-6-chloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-2-pyridylamine, 6-Chloropyridin-2-amine

L19836-03 L19836-06 L19836-14
CAAAAL19836-03EA 63.52 CAD
CAAAAL19836-03 CAAAAL19836-06 CAAAAL19836-14
2-Amino-6-chloropyridine 98%
2-Amino-6-chloropyridine

Purity: 98% CAS No.: 45644-21-1 Molecular Formula: C5H5ClN2 Formula Weight: 128.56 Melting Point: 69-72° Beilstein Reference: 108669

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00234068
CAS Number: 45644-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR