2-Fluoro-4-nitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19770-03
L19770-06
CAAAAL19770-03EA
142.04
CAD
CAAAAL19770-03
CAAAAL19770-06
2-Fluoro-4-nitrobenzoic acid 98%
2-Fluoro-4-nitrobenzoic acid
Purity: 98%
CAS No.: 403-24-7
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Melting Point: 172-175°
Beilstein Reference: 2582091
Formula:
C₇H₄FNO₄ MW: 185.11 g/mol Melting Pt: 172…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00275565 CAS Number: 403-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...