3-Cyano-4-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile

L19769-03 L19769-06
CAAAAL19769-03EA 70.47 CAD
CAAAAL19769-03 CAAAAL19769-06 CAAAAL19769-14
3-Cyano-4-methylpyridine 97%
3-Cyano-4-methylpyridine

Purity: 97% CAS No.: 5444-01-9 Molecular Formula: C7H6N2 Formula Weight: 118.14 Odor: Characteristic Melting Point: 41-44° Boiling Point: 108-111°/20mm Beilstein Reference: 112016 MDL No.: MFCD00234272

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 108…111 °C/20 mm
Melting Pt: 41…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00234272
CAS Number: 5444-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR