1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Butyl-3-methylimidazolium Trifluoromethanesulfonate, 1-Butyl-3-methylimidazolium Trifluoromethanesulphonate

L19765-04 L19765-09
CAAAAL19765-04EA 94.01 CAD
CAAAAL19765-04 CAAAAL19765-09
1-Butyl-3-methylimidazoliumtrifluoromethanesulfonate 98%
1-Butyl-3-methylimidazolium trifluoromethanesulphonate

Purity: 98% CAS No.: 174899-66-2 Molecular Formula: C9H15F3N2O3S Formula Weight: 288.29 Flash Point: None Refractive Index: 1.4390 Melting Point: 16° MDL No.: MFCD03427620

Formula: C₉H₁₅F₃N₂O₃S
MW: 288.29 g/mol
Melting Pt: 16 °C
Density: 1.295
Storage Temperature: Ambient
MDL Number: MFCD03427620
CAS Number: 174899-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR