1-Ethyl-3-methylimidazoliumtetrafluoroborate, (max. 3% H₂O) 98+% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate

L19763-04 L19763-09 L19763-18
CAAAAL19763-04EA 100.49 CAD
CAAAAL19763-04 CAAAAL19763-09 CAAAAL19763-18
1-Ethyl-3-methylimidazoliumtetrafluoroborate, (max. 3% H₂O) 98+% (dry weight)
1-Ethyl-3-methylimidazolium tetrafluoroborate

Purity: 98+% CAS No.: 143314-16-3 Molecular Formula: C6H11BF4N2 Formula Weight: 197.98 Flash Point: None Refractive Index: 1.4100 Melting Point: 15° MDL No.: MFCD00216668

Formula: C₆H₁₁BF₄N₂
MW: 197.97 g/mol
Melting Pt: 15 °C
Density: 1.280
Storage Temperature: Ambient
MDL Number: MFCD00216668
CAS Number: 143314-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR