1-Ethyl-3-methylimidazoliumhexafluorophosphate 98+%

Supplier: Thermo Scientific Chemicals
Warning

L19762-04 L19762-09 L19762-18
CAAAAL19762-04EA 102.38 CAD
CAAAAL19762-04 CAAAAL19762-09 CAAAAL19762-18
1-Ethyl-3-methylimidazoliumhexafluorophosphate 98+%
1-Ethyl-3-methylimidazolium hexafluorophosphate

Purity: 98+% CAS No.: 155371-19-0 Molecular Formula: C6H11F6N2P Formula Weight: 256.13 Melting Point: 58-62° MDL No.: MFCD00216703

Formula: C₆H₁₁F₆N₂P
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00216703
CAS Number: 155371-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR