1-Ethyl-3-methylimidazolium bromide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19761-09
L19761-18
CAAAAL19761-09EA
96.38
CAD
CAAAAL19761-09
CAAAAL19761-18
1-Ethyl-3-methylimidazolium bromide 98+%
1-Ethyl-3-methylimidazolium bromide
Purity: 98+%
CAS No.: 65039-08-9
Molecular Formula: C6H11BrN2
Formula Weight: 191.07
Melting Point: 70-73°
MDL No.: MFCD03427610
Formula:
C₆H₁₁BrN₂ Melting Pt: 70…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427610 CAS Number: 65039-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...