1,2,3,4-Tetramethylbenzene 95%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19756-06
L19756-14
CAAAAL19756-06EA
199.08
CAD
CAAAAL19756-06
CAAAAL19756-14
1,2,3,4-Tetramethylbenzene 95%, Technical Grade
1,2,3,4-Tetramethylbenzene
Purity: 95%
CAS No.: 488-23-3
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 68°(154°F)
Refractive Index: 1.5190
Boiling Point: 203°
Beilstein Reference: 1904390
MDL No.: MFCD00008521
Formula:
C₁₀H₁₄ Boiling Pt: 203 °C Melting Pt: –6 °C Density: 0.900 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008521 CAS Number: 488-23-3 EINECS: 207-673-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...