1-Butyl-3-methylimidazolium chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19749-09
L19749-18
CAAAAL19749-09EA
80.74
CAD
CAAAAL19749-09
CAAAAL19749-18
1-Butyl-3-methylimidazolium chloride 96%
1-Butyl-3-methylimidazolium chloride
Purity: 96%
CAS No.: 79917-90-1
Molecular Formula: C8H15ClN2
Formula Weight: 174.67
Melting Point: ca 73°
MDL No.: MFCD03095425
Formula:
C₈H₁₅ClN₂ Boiling Pt: ? 200 °C (1013 hPa) Melting Pt: ∼ 73 °C Density: ~ 1 g/cm³ (20 °C) Flash Pt: ≥ 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095425 CAS Number: 79917-90-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...