3-Amino-4-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19730-06
L19730-14
CAAAAL19730-06EA
114.71
CAD
CAAAAL19730-06
CAAAAL19730-14
3-Amino-4-methylpyridine 98%
3-Amino-4-methylpyridine
Purity: 98%
CAS No.: 3430-27-1
Molecular Formula: C6H8N2
Formula Weight: 108.14
Odor: Unpleasant
Melting Point: 102-106°
Boiling Point: 254°
Beilstein Reference: 107792
UN No.: 2811
MDL No.: MFCD00128871
Formula:
C₆H₈N₂ Boiling Pt: 254 °C Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128871 CAS Number: 3430-27-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...