3,5-Difluorobenzenesulfonamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19720-03
L19720-06
CAAAAL19720-03EA
68.1
CAD
CAAAAL19720-03
CAAAAL19720-06
3,5-Difluorobenzenesulfonamide 98%
3,5-Difluorobenzenesulphonamide
Purity: 98%
CAS No.: 140480-89-3
Molecular Formula: C6H5F2NO2S
Formula Weight: 193.17
Melting Point: 157-161°
Beilstein Reference: 5333160
Formula:
C₆H₅F₂NO₂S Melting Pt: 157…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD02091379 CAS Number: 140480-89-3 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...