2-Bromo-4-fluorobenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

L19713-03 L19713-06
CAAAAL19713-03EA 45.35 CAD
CAAAAL19713-03 CAAAAL19713-06
2-Bromo-4-fluorobenzenesulfonyl chloride 97%
2-Bromo-4-fluorobenzenesulphonyl chloride

Purity: 97% CAS No.: 351003-45-7 Molecular Formula: C6H3BrClFO2S Formula Weight: 273.51 Refractive Index: 1.5780 Boiling Point: 264-265° UN No.: 2920 MDL No.: MFCD03094149

Formula: C₆H₃BrClFO₂S
MW: 273.51 g/mol
Boiling Pt: 264…265 °C
Density: 1.870
Storage Temperature: Ambient
MDL Number: MFCD03094149
CAS Number: 351003-45-7
UN: 2920
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR