3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19711-03
L19711-06
CAAAAL19711-03EA
99.86
CAD
CAAAAL19711-03
CAAAAL19711-06
3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride 97%
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride
Purity: 97%
CAS No.: 351003-46-8
Molecular Formula: C7H3BrClF3O2S
Formula Weight: 323.52
Flash Point: 66°(150°F)
Refractive Index: 1.5240
Boiling Point: 217-218°
UN No.: 3265
MDL No.: MFCD03094393
Formula:
C₇H₃BrClF₃O₂S Boiling Pt: 217…218 °C Density: 1.835 Flash Pt: 66 °C (150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03094393 CAS Number: 351003-46-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...