4-Fluoro-m-anisaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19671-03
L19671-06
CAAAAL19671-03EA
63.67
CAD
CAAAAL19671-03
CAAAAL19671-06
4-Fluoro-m-anisaldehyde 98%
4-Fluoro-m-anisaldehyde
Purity: 98%
CAS No.: 128495-46-5
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Melting Point: 61-63°
Beilstein Reference: 5178063
Formula:
C₈H₇FO₂ Melting Pt: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143320 CAS Number: 128495-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...