5-Iodo-2-furaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19670-03
L19670-06
CAAAAL19670-03EA
72.21
CAD
CAAAAL19670-03
CAAAAL19670-06
5-Iodo-2-furaldehyde 97%
5-Iodo-2-furaldehyde
Purity: 97%
CAS No.: 2689-65-8
Molecular Formula: C5H3IO2
Formula Weight: 221.98
Storage Temperature: 2°C to 8°C
Melting Point: 126-127°
Beilstein Reference: 108406
MDL No.: MFCD00159503
Formula:
C₅H₃IO₂ Melting Pt: 126…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00159503 CAS Number: 2689-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...