5-Quinolinecarboxylic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19643-03
L19643-06
L19643-MD
CAAAAL19643-03EA
140.62
CAD
CAAAAL19643-03
CAAAAL19643-06
CAAAAL19643-MD
5-Quinolinecarboxylic acid 97+%
5-Quinolinecarboxylic acid
Purity: 97+%
CAS No.: 7250-53-5
Molecular Formula: C10H7NO2
Formula Weight: 173.17
Melting Point:
Formula:
C₁₀H₇NO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047608 CAS Number: 7250-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...