8-Quinolinecarbaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinoline-8-carbaldehyde, 8-Formylquinoline, 8-Quinolinecarboxaldehyde, Quinoline-8-carboxaldehyde

L19642-03 L19642-06
CAAAAL19642-03EA 210.14 CAD
CAAAAL19642-03 CAAAAL19642-06 CAAAAL19642-MD
8-Quinolinecarbaldehyde 98+%
8-Quinolinecarbaldehyde

Purity: 98+% CAS No.: 38707-70-9 Molecular Formula: C10H7NO Formula Weight: 157.17 Melting Point: 92-94° Beilstein Reference: 113039

Formula: C₁₀H₇NO
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00805837
CAS Number: 38707-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR