5-Quinolinecarbaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

L19641-03 L19641-06 L19641-MD
CAAAAL19641-03EA 285.98 CAD
CAAAAL19641-03 CAAAAL19641-06 CAAAAL19641-MD
5-Quinolinecarbaldehyde 97+%
5-Quinolinecarbaldehyde

Purity: 97+% CAS No.: 22934-41-4 Molecular Formula: C10H7NO Formula Weight: 157.17 Melting Point: 94-98° Beilstein Reference: 113095

Formula: C₁₀H₇NO
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00805835
CAS Number: 22934-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR