3-Cyanophenylboronic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(3-Cyanophenyl)metaboric acid, Dihydroxy(3-cyanophenyl)borane, 3-Cyanobenzeneboronic acid
L19635-03
L19635-06
CAAAAL19635-03EA
87.53
CAD
CAAAAL19635-03
CAAAAL19635-06
CAAAAL19635-MD
3-Cyanophenylboronic acid 98+%
3-Cyanophenylboronic acid
Purity: 98+%
CAS No.: 150255-96-2
Molecular Formula: C7H6BNO2
Formula Weight: 146.94
Melting Point: ca 298° dec.
Beilstein Reference: 6136720
Formula:
C₇H₆BNO₂ MW: 146.94 g/mol Melting Pt: ∼ 298 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01318967 CAS Number: 150255-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...