5-Aminoquinoline 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinolin-5-amine, Quinolin-5-ylamine

L19628-03 L19628-06
CAAAAL19628-03EA 85.32 CAD
CAAAAL19628-03 CAAAAL19628-06
5-Aminoquinoline 99%
5-Aminoquinoline

Purity: 99% CAS No.: 611-34-7 Molecular Formula: C9H8N2 Formula Weight: 144.18 Melting Point: 107-109° Boiling Point: 310° MDL No.: MFCD00006797

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 310 °C
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00006797
CAS Number: 611-34-7
EINECS: 210-266-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR