4-Methoxypyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19625-06
L19625-14
CAAAAL19625-06EA
124.5
CAD
CAAAAL19625-06
CAAAAL19625-14
4-Methoxypyridine 98+%
4-Methoxypyridine
Purity: 98+%
CAS No.: 620-08-6
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 74°(165°F)
Refractive Index: 1.5210
Melting Point: 4°
Boiling Point: 191°
Beilstein Reference: 108211
MDL No.: MFCD00674049
Formula:
C₆H₇NO Boiling Pt: 191 °C Melting Pt: 4 °C Density: 1.075 Flash Pt: 74 °C(165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00674049 CAS Number: 620-08-6 EINECS: 210-624-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...