2-Chloro-4-iodopyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

L19623-03 L19623-06
CAAAAL19623-03EA 63.52 CAD
CAAAAL19623-03 CAAAAL19623-06
2-Chloro-4-iodopyridine 99%
2-Chloro-4-iodopyridine

Purity: 99% CAS No.: 153034-86-7 Molecular Formula: C5H3ClIN Formula Weight: 239.44 Melting Point: 42-44° Beilstein Reference: 108668

Formula: C₅H₃ClIN
MW: 239.44
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD01861983
CAS Number: 153034-86-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR