3-Bromo-4-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

L19622-06 L19622-14
CAAAAL19622-03EA 131.28 CAD
CAAAAL19622-03 CAAAAL19622-06 CAAAAL19622-14
3-Bromo-4-methylpyridine 98%
3-Bromo-4-methylpyridine

Purity: 98% CAS No.: 3430-22-6 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 79°(174°F) Refractive Index: 1.5620 Boiling Point: 199-200° Beilstein Reference: 878354 MDL No.: MFCD00082592

Formula: C₆H₆BrN
Boiling Pt: 199…200 °C
Density: 1.549
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082592
CAS Number: 3430-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR