5-Bromo-2-iodopyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19610-06
L19610-14
CAAAAL19610-06EA
68.89
CAD
CAAAAL19610-06
CAAAAL19610-14
5-Bromo-2-iodopyridine 98%
5-Bromo-2-iodopyridine
Purity: 98%
CAS No.: 223463-13-6
Molecular Formula: C5H3BrIN
Formula Weight: 283.89
Melting Point: 113-116°
Beilstein Reference: 2119
Formula:
C₅H₃BrIN Melting Pt: 113…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00691506 CAS Number: 223463-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...