3-Chloro-2,5,6-trifluoropyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19543-06
L19543-14
CAAAAL19543-06EA
138.72
CAD
CAAAAL19543-06
CAAAAL19543-14
3-Chloro-2,5,6-trifluoropyridine 98+%
3-Chloro-2,5,6-trifluoropyridine
Purity: 98+%
CAS No.: 2879-42-7
Molecular Formula: C5HClF3N
Formula Weight: 167.52
Flash Point:
Formula:
C₅HClF₃N Boiling Pt: 132…134 °C Flash Pt: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03001160 CAS Number: 2879-42-7 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...