2,4,6-Trifluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

L19541-06 L19541-14
CAAAAL19541-06EA 229.1 CAD
CAAAAL19541-06 CAAAAL19541-14
2,4,6-Trifluoropyridine 97%
2,4,6-Trifluoropyridine

Purity: 97% CAS No.: 3512-17-2 Molecular Formula: C5H2F3N Formula Weight: 133.07 Flash Point: 17°(62°F) Refractive Index: 1.4130 Boiling Point: 102° Beilstein Reference: 1364338 UN No.: 2924 MDL No.: MFCD03001161

Formula: C₅H₂F₃N
Boiling Pt: 102 °C
Density: 1.382
Flash Pt: 17 °C (62 °F)
Storage Temperature: Ambient
MDL Number: MFCD03001161
CAS Number: 3512-17-2
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR