2,3,6-Trifluoropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19540-03
L19540-06
CAAAAL19540-03EA
66.36
CAD
CAAAAL19540-03
CAAAAL19540-06
2,3,6-Trifluoropyridine 97%
2,3,6-Trifluoropyridine
Purity: 97%
CAS No.: 3512-18-3
Molecular Formula: C5H2F3N
Formula Weight: 133.07
Flash Point: 30°(86°F)
Refractive Index: 1.4200
Boiling Point: 100-102°
Beilstein Reference: 1365146
UN No.: 1993
MDL No.: MFCD03001158
Formula:
C₅H₂F₃N Boiling Pt: 100…102 °C Density: 1.402 Flash Pt: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03001158 CAS Number: 3512-18-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...