5-Bromo-2-fluoropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19537-06
L19537-14
CAAAAL19537-06EA
162.74
CAD
CAAAAL19537-06
CAAAAL19537-14
5-Bromo-2-fluoropyridine 98%
5-Bromo-2-fluoropyridine
Purity: 98%
CAS No.: 766-11-0
Molecular Formula: C5H3BrFN
Formula Weight: 175.99
Flash Point: 73°(163°F)
Refractive Index: 1.5294
Odor: Characteristic
Boiling Point: 162-164°
Beilstein Reference: 1363171
UN No.: 2810
MDL No.: MFCD01863742
Formula:
C₅H₃BrFN MW: 175.99 g/mol Boiling Pt: 162…164 °C Density: 1.72 Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01863742 CAS Number: 766-11-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...