2-Phenylethylboronic acid pinacol ester 99%

Supplier: Thermo Scientific Chemicals

L19530-03 L19530-MD
CAAAAL19530-03EA 153.89 CAD
CAAAAL19530-03 CAAAAL19530-MD
2-Phenylethylboronic acid pinacol ester 99%
2-Phenylethylboronic acid pinacol ester

Purity: 99% CAS No.: 165904-22-3 Molecular Formula: C14H21BO2 Formula Weight: 232.13 Refractive Index: 1.4860 Melting Point: 31-33° Boiling Point: 70-80° Beilstein Reference: 8258927 MDL No.: MFCD03788721

Formula: C₁₄H₂₁BO₂
Boiling Pt: 70…80 °C
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD03788721
CAS Number: 165904-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR