2-Hydroxy-5-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
L19528-06
L19528-14
CAAAAL19528-06EA
107.91
CAD
CAAAAL19528-06
CAAAAL19528-14
2-Hydroxy-5-methylpyridine 98%
2-Hydroxy-5-methylpyridine
Purity: 98%
CAS No.: 1003-68-5
Molecular Formula: C6H7NO
Formula Weight: 109.13
Odor: Odorless
Melting Point: 183-187°
Beilstein Reference: 107074
MDL No.: MFCD03092888
Formula:
C₆H₇NO Melting Pt: 183…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD03092888 CAS Number: 1003-68-5 EINECS: 213-719-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...