3-Bromo-2-nitropyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

L19523-03 L19523-06
CAAAAL19523-03EA 84.53 CAD
CAAAAL19523-03 CAAAAL19523-06
3-Bromo-2-nitropyridine 98+%
3-Bromo-2-nitropyridine

Purity: 98+% CAS No.: 54231-33-3 Molecular Formula: C5H3BrN2O2 Formula Weight: 203.00 Melting Point: 102-106° Beilstein Reference: 1637071

Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
Melting Pt: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00955614
CAS Number: 54231-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR