4-Fluoro-2-methylbenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19517-03
L19517-06
CAAAAL19517-03EA
48.03
CAD
CAAAAL19517-03
CAAAAL19517-06
4-Fluoro-2-methylbenzenesulfonyl chloride 97%
4-Fluoro-2-methylbenzenesulphonyl chloride
Purity: 97%
CAS No.: 7079-48-3
Molecular Formula: C7H6ClFO2S
Formula Weight: 208.64
Flash Point:
Formula:
C₇H₆ClFO₂S Boiling Pt: 245…246 °C Density: 1.433 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03094390 CAS Number: 7079-48-3 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...