1,3-Difluoro-5-iodobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Difluoroiodobenzene
L19510-03
L19510-06
CAAAAL19510-03EA
61.62
CAD
CAAAAL19510-03
CAAAAL19510-06
1,3-Difluoro-5-iodobenzene 98%
1,3-Difluoro-5-iodobenzene
Purity: 98%
CAS No.: 2265-91-0
Molecular Formula: C6H3F2I
Formula Weight: 239.99
Flash Point: 82°(179°F)
Refractive Index: 1.5540
Boiling Point: 173-174°
Beilstein Reference: 2041185
MDL No.: MFCD03094155
Formula:
C₆H₃F₂I MW: 239.99 g/mol Boiling Pt: 173…174 °C Flash Pt: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03094155 CAS Number: 2265-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...