2,4,5-Trichloropyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

L19506-03 L19506-06
CAAAAL19506-03EA 67.31 CAD
CAAAAL19506-03 CAAAAL19506-06
2,4,5-Trichloropyrimidine 98%
2,4,5-Trichloropyrimidine

Purity: 98% CAS No.: 5750-76-5 Molecular Formula: C4HCl3N2 Formula Weight: 183.43 Flash Point: 110°(230°F) Refractive Index: 1.5730 Boiling Point: 84°/1mm Beilstein Reference: 4449 UN No.: 1760 MDL No.: MFCD03788200

Formula: C₄HCl₃N₂
MW: 183.42 g/mol
Boiling Pt: 84 °C/1 mm
Density: 1.600
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03788200
CAS Number: 5750-76-5
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR