2-Cyano-3-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19456-06
L19456-14
L19456-22
CAAAAL19456-06EA
67.31
CAD
CAAAAL19456-06
CAAAAL19456-14
CAAAAL19456-22
2-Cyano-3-methylpyridine 98%
2-Cyano-3-methylpyridine
Purity: 98%
CAS No.: 20970-75-6
Molecular Formula: C7H6N2
Formula Weight: 118.14
Flash Point: 115°(239°F)
Odor: Characteristic
Boiling Point: 141°/38mm
Beilstein Reference: 112001
MDL No.: MFCD00190585
Formula:
C₇H₆N₂ Boiling Pt: 141 °C/38 mm Melting Pt: 84…87 °C Flash Pt: 115 °C(239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190585 CAS Number: 20970-75-6 EINECS: 244-131-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...