2-Amino-6-(trifluoromethyl)pyridine 98+%

Supplier: Thermo Scientific Chemicals
Danger

L19443-MC L19443-ME
CAAAAL19443-MCEA 98.75 CAD
CAAAAL19443-MC CAAAAL19443-ME
2-Amino-6-(trifluoromethyl)pyridine 98+%
2-Amino-6-(trifluoromethyl)pyridine

Purity: 98+% CAS No.: 34486-24-3 Molecular Formula: C6H5F3N2 Formula Weight: 162.11 Melting Point: 86-89° Beilstein Reference: 473259 UN No.: 2811 MDL No.: MFCD03093868

Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03093868
CAS Number: 34486-24-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR