1,4-Benzenedithiol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19440-03
L19440-06
L19440-MD
CAAAAL19440-03EA
249.64
CAD
CAAAAL19440-03
CAAAAL19440-06
CAAAAL19440-MD
1,4-Benzenedithiol 97%
1,4-Benzenedithiol
Purity: 97%
CAS No.: 624-39-5
Molecular Formula: C6H6S2
Formula Weight: 142.24
Storage Temperature: 2°C to 8°C
Odor: Stench
Beilstein Reference: 1680022
UN No.: 3335
MDL No.: MFCD00142541
Formula:
C₆H₆S₂ Boiling Pt: 240 °C (1013 hPa) Melting Pt: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142541 CAS Number: 624-39-5 UN: 3335 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...