5-Bromo-2,3-diaminopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine

L19435-06 L19435-14
CAAAAL19435-06EA 27.33 CAD
CAAAAL19435-06 CAAAAL19435-14
5-Bromo-2,3-diaminopyridine 97%
5-Bromo-2,3-diaminopyridine

Purity: 97% CAS No.: 38875-53-5 Molecular Formula: C5H6BrN3 Formula Weight: 188.03 Melting Point: 153-161° Beilstein Reference: 119436 UN No.: 2811 MDL No.: MFCD00460094

Formula: C₅H₆BrN₃
MW: 188.03 g/mol
Boiling Pt: 321
Melting Pt: 153…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00460094
CAS Number: 38875-53-5
EINECS: 254-172-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR