3-Isoquinolinol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxyisoquinoline

L19428-03 L19428-06
CAAAAL19428-03EA 116.45 CAD
CAAAAL19428-03 CAAAAL19428-06
3-Isoquinolinol 99%
3-Isoquinolinol

Purity: 99% CAS No.: 7651-81-2 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: ca 250° dec. Beilstein Reference: 113230

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: ∼ 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075524
CAS Number: 7651-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR