1,3-Dimethoxy-4-iodobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Iodo-2,4-dimethoxybenzene, 2,4-Dimethoxyiodobenzene

L19420-03 L19420-06
CAAAAL19420-03EA 37.29 CAD
CAAAAL19420-03 CAAAAL19420-06
1,3-Dimethoxy-4-iodobenzene 97%
1,3-Dimethoxy-4-iodobenzene

Purity: 97% CAS No.: 20469-63-0 Molecular Formula: C8H9IO2 Formula Weight: 264.06 Flash Point: 110°(230°F) Refractive Index: 1.6100 Melting Point: 37-41° Boiling Point: 135°/14mm Beilstein Reference: 1945135 MDL No.: MFCD00059268

Formula: C₈H₉IO₂
MW: 264.06 g/mol
Boiling Pt: 135 °C/14 mm
Melting Pt: 37…41 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059268
CAS Number: 20469-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR