2,4,6-Trifluorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane

L19402-03 L19402-06
CAAAAL19402-03EA 67.31 CAD
CAAAAL19402-03 CAAAAL19402-06
2,4,6-Trifluorophenylboronic acid 97%
2,4,6-Trifluorophenylboronic acid

Purity: 97% CAS No.: 182482-25-3 Molecular Formula: C6H4BF3O2 Formula Weight: 175.90 Melting Point: ca 235° Beilstein Reference: 8549766

Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
Melting Pt: ∼ 235 °C
Storage Temperature: Ambient
MDL Number: MFCD01863169
CAS Number: 182482-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR