2-Hydroxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxybenzeneboronic acid, Dihydroxy(2-hydroxyphenyl)borane, (2-Hydroxyphenyl)metaboric acid
L19400-03
L19400-06
CAAAAL19400-03EA
64.46
CAD
CAAAAL19400-03
CAAAAL19400-06
2-Hydroxyphenylboronic acid 97%
2-Hydroxyphenylboronic acid
Purity: 97%
CAS No.: 89466-08-0
Molecular Formula: C6H7BO3
Formula Weight: 137.93
Melting Point: ca 185°
Beilstein Reference: 2962938
Formula:
C₆H₇BO₃ MW: 137.93 g/mol Melting Pt: ∼ 185 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074581 CAS Number: 89466-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...