1-(4-Pyridyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(1-Piperazinyl)pyridine, 1-(Pyridin-4-yl)piperazine

L19396-06 L19396-14
CAAAAL19396-06EA 221.2 CAD
CAAAAL19396-06 CAAAAL19396-14
1-(4-Pyridyl)piperazine 97%
1-(4-Pyridyl)piperazine

Purity: 97% CAS No.: 1008-91-9 Molecular Formula: C9H13N3 Formula Weight: 163.22 Melting Point: 136-141° Boiling Point: 195-200°/0.3mm UN No.: 3259 MDL No.: MFCD00040745

Formula: C₉H₁₃N₃
MW: 163.22 g/mol
Boiling Pt: 195…200 °C/0.3 mm
Melting Pt: 140
Storage Temperature: Ambient
MDL Number: MFCD00040745
CAS Number: 1008-91-9
EINECS: 213-764-7
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR