2-Bromo-4-fluorobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19321-06
L19321-14
CAAAAL19321-03EA
53.94
CAD
CAAAAL19321-03
CAAAAL19321-06
CAAAAL19321-14
2-Bromo-4-fluorobenzoic acid 98%
2-Bromo-4-fluorobenzoic acid
Purity: 98%
CAS No.: 1006-41-3
Molecular Formula: C7H4BrFO2
Formula Weight: 219.01
Melting Point: 167-169°
Beilstein Reference: 2614292
Formula:
C₇H₄BrFO₂ MW: 219.01 g/mol Boiling Pt: 289 Melting Pt: 167…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055370 CAS Number: 1006-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...