2'-Bromo-4'-fluoroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

L19320-03 L19320-06
CAAAAL19320-03EA 108.39 CAD
CAAAAL19320-03 CAAAAL19320-06
2'-Bromo-4'-fluoroacetophenone 98%
2'-Bromo-4'-fluoroacetophenone

Purity: 98% CAS No.: 1006-39-9 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Flash Point: 110°(230°F) Refractive Index: 1.5490 Boiling Point: 150°/12mm Beilstein Reference: 2249797 MDL No.: MFCD00077464

Formula: C₈H₆BrFO
Boiling Pt: 150 °C/12 mm
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077464
CAS Number: 1006-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR