2-Fluorobenzothiazole 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19316-03
L19316-06
L19316-MD
CAAAAL19316-03EA
169.06
CAD
CAAAAL19316-03
CAAAAL19316-06
CAAAAL19316-MD
2-Fluorobenzothiazole 99%
2-Fluorobenzothiazole
Purity: 99%
CAS No.: 1123-98-4
Molecular Formula: C7H4FNS
Formula Weight: 153.18
Refractive Index: 1.5830
Odor: Characteristic
Boiling Point: 97-98°/16mm
Beilstein Reference: 971503
UN No.: 2810
MDL No.: MFCD00047254
Formula:
C₇H₄FNS Boiling Pt: 97…98 °C/16 mm Density: 1.330 Storage Temperature: Ambient |
MDL Number:
MFCD00047254 CAS Number: 1123-98-4 EINECS: 214-384-4 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...