3-Amino-2-naphthoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-2-naphthoic Acid

L19312-03 L19312-06
CAAAAL19312-03EA 79 CAD
CAAAAL19312-03 CAAAAL19312-06
3-Amino-2-naphthoic acid 97%
3-Amino-2-naphthoic acid

Purity: 97% CAS No.: 5959-52-4 Molecular Formula: C11H9NO2 Formula Weight: 187.20 Melting Point: ca 210° dec. Beilstein Reference: 744099 Merck Reference: 14,452 MDL No.: MFCD00004115

Formula: C₁₁H₉NO₂
Boiling Pt: 383 °C (1013 hPa)
Melting Pt: ∼ 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004115
CAS Number: 5959-52-4
EINECS: 227-726-2
Merck Index: 14,452

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR