(R)-(+)-1-Phenylethanol ≥99%, ee 97%, ChiPros®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-1-Phenylethanol, (R)-(+)-α-Methylbenzyl alcohol, (R)-(+)-sec-Phenethyl alcohol
L19296-06
L19296-14
CAAAAL19296-06EA
197.5
CAD
CAAAAL19296-06
CAAAAL19296-14
(R)-(+)-1-Phenylethanol ≥99%, ee 97%, ChiPros®
(R)-(+)-1-Phenylethanol
For synthesis of optically active products
Purity: 99%
CAS No.: 1517-69-7
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 85°(185°F)
Refractive Index: 1.5280
Odor: Mild
Melting Point: 9-11°
Boiling Point: 88-89°/10mm
Beilstein Reference: 2039798
Optical Rotation: +45° (c=5 in methanol)
UN No.: 2937
MDL No.: MFCD00064263
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 88…89 °C/10 mm Melting Pt: 9…11 °C Density: 1.012 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064263 CAS Number: 1517-69-7 EINECS: 604-814-2 UN: 2937 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...